missing translation for 'onlineSavingsMsg'
Learn More

PSI-6206 13C,d3, MedChemExpress

Product Code. 16436388
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16436388

Brand: Medchem Express HY15236S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.

PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.

Specifications

Chemical Name or Material PSI-6206 13C,d3
CAS 1256490-42-2
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C9 13CH10D3FN2O5
Color Light Yellow
Synonym RO-2433 13C,d3 GS-331007 13C,d3 Sofosbuvir metabolite GS-331007 13C,d3
SMILES OC[C@@H]1[C@H]([C@@]([13C]([2H])([2H])[2H])(F)[C@H](N2C(NC(C=C2)=O)=O)O1)O
Molecular Weight (g/mol) 264.23
Quantity 10 mg
Formula Weight 264.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.