missing translation for 'onlineSavingsMsg'
Learn More

PSI-6130, MedChemExpress

Product Code. 16469956
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469956 50 mg 50mg
16460056 2 mg 2mg
16456207 5 mg 5mg
16403488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469956 Supplier Medchem Express Supplier No. HY10165
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.

PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.

Specifications

Chemical Name or Material PSI-6130
CAS 817204-33-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H14FN3O4
Quantity 50 mg
Synonym R 1656
Solubility Information DMSO : 25 mg/mL (96.44 mM; Need ultrasonic)
SMILES O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2O)F)O[C@@H]2CO)N
Molecular Weight (g/mol) 259.23
Formula Weight 259.23
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.