missing translation for 'onlineSavingsMsg'
Learn More

PSGL-1/CD162 Antibody (PSGL1/1567) - Azide and BSA Free, Novus Biologicals™

Product Code. 18374012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374012 Supplier Novus Biologicals Supplier No. NBP308695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSGL-1/CD162
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PSGL1/1567
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA
Host Species Mouse
Immunogen Recombinant human PSGL-1/CD162 protein
Purification Method Column Chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6404
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.