missing translation for 'onlineSavingsMsg'
Learn More

PSGL-1/CD162 Antibody (688102) [DyLight 650], Novus Biologicals™

Product Code. 30732565 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732565 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732565 Supplier Novus Biologicals Supplier No. FAB3345W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSGL-1/CD162 Monoclonal antibody specifically detects PSGL-1/CD162 in Human samples. It is validated for Immunohistochemistry,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSGL-1/CD162
Applications Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 688102
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PSGL-1, Gln42-Gly295, Accession # Q14242
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6404
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.