missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSG6 Monoclonal Antibody (935981)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561968
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The human pregnancy-specific glycoproteins (PSGs) are a group of molecules that are mainly produced by the placental syncytiotrophoblasts during pregnancy. PSGs comprise a subgroup of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily. For additional general information about the PSG gene family, see PSG1 (MIM 176390).
Specifications
| PSG6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q00889 | |
| PSG6 | |
| Human embryonic kidney cell line HEK-293-derived transfected with human PSG6, Met1-Gly424. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 935981 | |
| Unconjugated | |
| PSG6 | |
| CGM3; pregnancy specific beta-1-glycoprotein 6; pregnancy-specific beta-1-glycoprotein 10; Pregnancy-specific beta-1-glycoprotein 12; pregnancy-specific beta-1-glycoprotein 6; PS-beta-G-10; PS-beta-G-12; PS-beta-G-6; PSBG-10; PSBG-12; PSBG-6; PSG10; PSG12; PSG6; PSGGB | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction