missing translation for 'onlineSavingsMsg'
Learn More

PSG6 Antibody (935981) [DyLight 650], Novus Biologicals™

Product Code. 30668573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30668573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30668573 Supplier Novus Biologicals Supplier No. FAB85982W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSG6 Monoclonal antibody specifically detects PSG6 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG6
Applications ELISA
Classification Monoclonal
Clone 935981
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CGM3, pregnancy specific beta-1-glycoprotein 6, Pregnancy-specific beta-1-glycoprotein 10, Pregnancy-specific beta-1-glycoprotein 12, pregnancy-specific beta-1-glycoprotein 6, Pregnancy-specific glycoprotein 10, Pregnancy-specific glycoprotein 12, Pregnancy-specific glycoprotein 6, PS-beta-G-10, PS-beta-G-12, PS-beta-G-6, PSBG-10, PSBG-12, PSBG-6, PSG10, PSG12, PSGGB
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK-293-derived transfected with human PSG6, Met1-Gly424, Accession # Q00889
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5675
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.