missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95167-0.1ml
This item is not returnable.
View return policy
Description
PSG2 Polyclonal antibody specifically detects PSG2 in Human samples. It is validated for Western Blot
Specifications
| PSG2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| carcinoembryonic antigen SG8, pregnancy specific beta-1-glycoprotein 2, Pregnancy-specific beta-1 glycoprotein E, pregnancy-specific beta-1-glycoprotein 2, pregnancy-specific beta-1-glycoprotein 7, pregnancy-specific beta-1-glycoprotein-2, Pregnancy-specific glycoprotein 2, PS-beta-E, PS-beta-G-2, PSBG-2, PSBG2CEA, PSG1, PSGGB | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human PSG2 (NP_112536.2). RTLFLFGVTKYTAGPYECEIRNSGSASRSDPVTLNLLHGPDLPRIHPSYTNYRSGDNLYLSCFANSNPPAQYSWTINGKFQQSGQNLFIPQITTKHSGLYV | |
| 0.1 mL | |
| Cell Biology | |
| 5670 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction