missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11809-1-AP-150UL
This item is not returnable.
View return policy
Description
Human pregcy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregcy thus protecting the semiallotypic fetus from rejection.Specifications
| PSG1 | |
| Polyclonal | |
| Unconjugated | |
| PSG1 | |
| B1G1, CD66f, DHFRP2, FL NCA 1/2, PBG1, PS beta C/D, PS beta G 1, PSBG 1, PSBG1, PSG1, PSG95, PSGGA, PSGIIA, SP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5669 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11464 | |
| PSG1 | |
| PSG1 Fusion Protein Ag2468 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction