missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG1 Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90307-100UL
This item is not returnable.
View return policy
Description
PSG1 Monoclonal specifically detects PSG1 in Human samples. It is validated for ELISA.
Specifications
| PSG1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5669 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 2 | |
| ELISA 1:5000-1:10000 | |
| B1G1CD66 antigen-like family member F, CD66f, CD66f antigen, DHFRP2, Fetal liver non-specific cross-reactive antigen 1/2, FLJ90598, FLJ90654, FL-NCA-1/2, PBG1, pregnancy specific beta-1-glycoprotein 1, pregnancy-specific B-1 glycoprotein, Pregnancy-specific beta-1 glycoprotein C/D, pregnancy-specific beta-1-glycoprotein 1, Pregnancy-specific glycoprotein 1, PS-beta-C/D, PS-beta-G-1, PSBG-1, PSBG1SP1, PSGGAPSG95, PSGIIA | |
| Recombinant Human PSG1 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction