missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG1 Antibody (002), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90307CL1
This item is not returnable.
View return policy
Description
PSG1 Monoclonal antibody specifically detects PSG1 in Human samples. It is validated for ELISA
Specifications
| PSG1 | |
| Monoclonal | |
| CoraFluor 1 | |
| B1G1CD66 antigen-like family member F, CD66f, CD66f antigen, DHFRP2, Fetal liver non-specific cross-reactive antigen 1/2, FLJ90598, FLJ90654, FL-NCA-1/2, PBG1, pregnancy specific beta-1-glycoprotein 1, pregnancy-specific B-1 glycoprotein, Pregnancy-specific beta-1 glycoprotein C/D, pregnancy-specific beta-1-glycoprotein 1, Pregnancy-specific glycoprotein 1, PS-beta-C/D, PS-beta-G-1, PSBG-1, PSBG1SP1, PSGGAPSG95, PSGIIA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PSG1 (Accession#: NP_001171754.1; Met1-Pro419). | |
| 0.1 mL | |
| CD Markers, Immunology | |
| 5669 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 002 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction