missing translation for 'onlineSavingsMsg'
Learn More

PSF3 Antibody (OTI3A12), DyLight 594, Novus Biologicals™

Product Code. 18413046 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413046 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413046 Supplier Novus Biologicals Supplier No. NBP273680DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSF3 Monoclonal antibody specifically detects PSF3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSF3
Applications Western Blot
Classification Monoclonal
Clone OTI3A12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DNA replication complex GINS protein PSF3, GINS complex subunit 3, GINS complex subunit 3 (Psf3 homolog), PSF3FLJ13912
Host Species Mouse
Immunogen Full length recombinant protein of human PSF3 (NP_073607) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64785
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.