missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38597-100ul
This item is not returnable.
View return policy
Description
PSF2 Polyclonal antibody specifically detects PSF2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| PSF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DNA replication complex GINS protein PSF2, GINS complex subunit 2, GINS complex subunit 2 (Psf2 homolog), HSPC037, PSF2Pfs2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human PSF2 (NP_057179.1).,, Sequence:, MDAAEVEFLAEKELVTIIPNFSLDKIYLIGGDLGPFNPGLPVEVPLWLAINLKQRQKCRLLPPEWMDVEKLEKMRDHERKEETFTPMPSPYYMELTKLLLNHASDNIPKADEIRTLVKDMWDTRIAKLRVSADSFVRQQEAHAKLDNLTLMEINTSGTFLTQALNHMYKLRTNLQPLESTQSQDF | |
| 100 μL | |
| Cell Cycle and Replication | |
| 51659 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction