missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | PSF2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PSF2 Polyclonal specifically detects PSF2 in Human samples. It is validated for Western Blot.Specifications
| PSF2 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| DNA replication complex GINS protein PSF2, GINS complex subunit 2, GINS complex subunit 2 (Psf2 homolog), HSPC037, PSF2Pfs2 | |
| GINS2 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9Y248 | |
| 51659 | |
| Synthetic peptides corresponding to GINS2(GINS complex subunit 2 (Psf2 homolog)) The peptide sequence was selected from the C terminal of GINS2. Peptide sequence VRQQEAHAKLDNLTLMEINTSGTFLTQALNHMYKLRTNLQPLESTQSQDF. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title