missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PSF2 Monoclonal antibody specifically detects PSF2 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
Specifications
Specifications
| Antigen | PSF2 |
| Applications | ELISA, Western Blot, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 3H4 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH03186 |
| Gene Alias | DNA replication complex GINS protein PSF2, GINS complex subunit 2, GINS complex subunit 2 (Psf2 homolog), HSPC037, PSF2Pfs2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?