missing translation for 'onlineSavingsMsg'
Learn More

PSF1 Antibody, Biotin, Novus Biologicals™

Product Code. 18423862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423862 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423862 Supplier Novus Biologicals Supplier No. NBP232156B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSF1 Polyclonal antibody specifically detects PSF1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSF1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias GINS complex subunit 1, GINS complex subunit 1 (Psf1 homolog), KIAA0186PSF1DNA replication complex GINS protein PSF1, partner of sld five-1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 146 to 196 of human GINS Complex Subunit 1 using the numbering given in entry NP_066545.3 (GeneID 9837).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9837
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.