missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32156AF647
This item is not returnable.
View return policy
Description
PSF1 Polyclonal antibody specifically detects PSF1 in Human samples. It is validated for Western Blot
Specifications
| PSF1 | |
| Polyclonal | |
| Western Blot | |
| GINS complex subunit 1, GINS complex subunit 1 (Psf1 homolog), KIAA0186PSF1DNA replication complex GINS protein PSF1, partner of sld five-1 | |
| The immunogen this antibody was made to, maps to a region between residue 146 to 196 of human GINS Complex Subunit 1 using the numbering given in entry NP_066545.3 (GeneID 9837). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9837 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction