missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF™ Oxford Safety Shoes
Specifications
Specifications
| Color | Black |
| Material | Leather |
| Size | 11 |
| Sole Features | Antistatic dual density sole |
| Sole Material | Polyurethane |
| Style | Oxford |
| Toe Material | Steel |
| Toe Style | Cap |
| Resistant To | Oils, Acids, Alkalis |
| Product Type | Oxford Safety Shoe |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?