Learn More
PSF™ Leather 338SM Model Shoes
Product Code. 12862955
Click to view available options
Size:
3
4
5
6
7
Product Code. 12862955
Brand: PSF Progressive Safety 338SM3
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Color Black
Material Leather
Size 3
Sole Features Lightweight
Sole Material Polyurethane
Weight (Metric) 1 kg
Resistant To Oils, chemicals and heat
Product Type Shoes