missing translation for 'onlineSavingsMsg'
Learn More

PSF™ Contractor Safety Shoes

Product Code. p-8008498
Click to view available options
Size:
6
7
8
9
10
13
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 12857583

Brand: PSF™ 806SM9

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance S1 Rated
Color Black
For Use With (Application) Industry, Maintenance, College, Mechanics
Material Leather
Size 9
Sole Features Dual Density
Sole Material Polyurethane
Style Contractor Safety
Toe Material Steel
Resistant To Heat, oils and general chemicals
Product Type Contractor Safety Shoe
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.