missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (1010/287), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45176F
This item is not returnable.
View return policy
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Pseudomonas Aeruginosa | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| AERUGINOSA | |
| Store at 4C in the dark. | |
| IgG2a |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 1010/287 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Pseudomonas aeruginosa 9 | |
| P. aeruginosa serotype 6 | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction