missing translation for 'onlineSavingsMsg'
Learn More

Pseudoerythromycin A Enol Ether, Mikromol™

Product Code. 30051381
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051381 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051381 Supplier Mikromol Supplier No. MM0132.110025
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pseudoerythromycin A Enol Ether
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pseudo Erythromycin A Enol Ether
CAS 105882-69-7
Analyte or Component Names Pseudo Erythromycin A Enol Ether
Shipping Condition Ice pack (-20°C)
Molecular Formula C37 H65 N O12
Certifications/Compliance ISO 9001
Synonym 4,13-Dioxabicyclo[8.2.1]tridec-12-en-5-one, 7-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-[(1R,2R)-1,2-dihydroxy-1-methylbutyl]-2,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, (2R,3R,6R,7S,8S,9R,10R)-, 4,13-Dioxabicyclo[8.2.1]tridec-12-en-5-one, 7-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-(1,2-dihydroxy-1-methylbutyl)-2,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, [2R-[2R*,3R*(1R*,2R*),6R*,7S*,8S*,9R*,10R*]]-, (2R,3R,6R,7S,8S,9R,10R)-7-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-[(1R,2R)-1,2-dihydroxy-1-methylbutyl]-2,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-4,13-dioxabicyclo[8.2.1]tridec-12-en-5-one, 8,9-Anhydropseudo-erythromycin A-6,9-hemiketal, EM 701, LY 267108, Pseudoerythromycin A enol ether
InChI Formula InChI=1S/C37H65NO12/c1-14-25(39)37(10,43)32-20(4)28-18(2)16-36(9,50-28)31(49-34-27(40)24(38(11)12)15-19(3)45-34)21(5)29(22(6)33(42)48-32)47-26-17-35(8,44-13)30(41)23(7)46-26/h19-27,29-32,34,39-41,43H,14-17H2,1-13H3/t19-,20+,21+,22-,23+,24+,25-,26+,27-,29+,30+,31-,32-,34+,35-,36-,37-/m1/s1
SMILES CC[C@@H](O)[C@@](C)(O)[C@@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)CC(=C(O4)[C@@H]1C)C
IUPAC Name (2R,3R,6R,7S,8S,9R,10R)-3-[(2R,3R)-2,3-dihydroxypentan-2-yl]-9-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-7-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-2,6,8,10,12-pentamethyl-4,13-dioxabicyclo[8.2.1]tridec-1(12)-en-5-one
Molecular Weight (g/mol) 715.91
Quantity 25 mg
Formula Weight 715.4507 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.