missing translation for 'onlineSavingsMsg'
Learn More

PSENEN/PEN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NB300-227SS

 View more versions of this product

Product Code. 18243650

  • £171.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PSENEN/PEN2
Polyclonal
Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100
Q9NZ42
PSENEN
A synthetic peptide corresponding to the C-terminus of human PEN2 (within residues 50-101). [UniProt# Q9NZ42]
Unpurified
RUO
Primary
Human and mouse neuroblastoma (N2a) cells.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation
Unconjugated
Whole antisera with 0.05% Sodium Azide
gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2
Rabbit
12 kDa
0.025 mL
Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
55851
Human, Mouse
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only