missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSEN2 Polyclonal Antibody

Brand: Thermo Scientific PA5112675
This item is not returnable.
View return policy
Description
Specifications
| PSEN2 | |
| Polyclonal | |
| Unconjugated | |
| PSEN2 | |
| AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5664, 81751 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O88777, P49810 | |
| PSEN2 | |
| Recombinant Human Presenilin-2 protein (7-77AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction