missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD93 Antibody (S18-30), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12912AF594
This item is not returnable.
View return policy
Description
PSD93 Monoclonal antibody specifically detects PSD93 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PSD93 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S18-30 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Chapsyn-110, discs, large homolog 2 (Drosophila), discs, large homolog 2, chapsyn-110, discs, large homolog 2, chapsyn-110 (Drosophila), disks large homolog 2, DKFZp781D1854, DKFZp781E0954, FLJ37266, MGC131811, Postsynaptic density protein PSD-93, PSD-93, PSD93,110kDa | |
| Fusion protein amino acids 1-852 of rat Chapsyn-110 | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 1740 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction