missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17298-100UL
This item is not returnable.
View return policy
Description
PSD4 Polyclonal antibody specifically detects PSD4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PSD4 | |
| Polyclonal | |
| Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| pleckstrin and Sec7 domain containing 4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: HSSGILPKWTLDASQSSLLETDGEQPSSLKKKEAGEAPKPGEEVKSEGTARPAETGDVQPDIHLTSAEHENLRTPMNSSWLPG | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23550 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction