missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557030
This item is not returnable.
View return policy
Description
Immunogen sequence: PGSPVNSHLP GSPKQNRSTS TQVVFWAGIL QAQMCVLDLE EELEKTEGLK AGLKCCLPTP PVDLPGDTGL HSSPPENEDS GEDSSEPEGE GQAWLREGTP DSSPQW Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 67%.
Guanine nucleotide exchange factor for ARF6 and ARL14/ARF7. Through ARL14 activation, controls the movement of MHC class II-containing vesicles along the actin cytoskeleton in dendritic cells. Involved in membrane recycling. Interacts with several phosphatidylinositol phosphate species, including phosphatidylinositol 3,4-bisphosphate, phosphatidylinositol 3,5-bisphosphate and phosphatidylinositol 4,5-bisphosphate. [UniProt].
Specifications
| PSD4 | |
| Polyclonal | |
| Unconjugated | |
| PSD4 | |
| ADP-ribosylation factor guanine nucleotide-exchange factor 6; EFA6B; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 B; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6B; exchange factor for ARF6 B; PH and SEC7 domain-containing protein 4; pleckstrin and Sec7 domain containing 4; pleckstrin homology and SEC7 domain-containing protein 4; PSD4; SEC7 homolog; telomeric of interleukin-1 cluster protein; TIC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23550 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NDX1 | |
| PSD4 | |
| Recombinant protein corresponding to Human PSD4. Recombinant protein control fragment (Product #RP-102631). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction