missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18437UV
This item is not returnable.
View return policy
Description
PSD3 Polyclonal antibody specifically detects PSD3 in Human samples. It is validated for Western Blot
Specifications
| PSD3 | |
| Polyclonal | |
| Western Blot | |
| ADP-ribosylation factor guanine nucleotide factor 6, DKFZp761K1423, EFA6RHepatocellular carcinoma-associated antigen 67, Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6, HCA67Pleckstrin homology and SEC7 domain-containing protein 3, KIAA0942, PH and SEC7 domain-containing protein 3, pleckstrin and Sec7 domain containing 3 | |
| Recognizes region between residues Between Between 998 and 1048.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23362 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction