missing translation for 'onlineSavingsMsg'
Learn More

PSD2 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30761620 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761620 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761620 Supplier Novus Biologicals Supplier No. NBP346870CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSD2 Polyclonal antibody specifically detects PSD2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias DKFZp761B0514, EC 2.5.1.6, EC 3.6.1,6330404E20Rik, PH and SEC7 domain-containing protein 2, pleckstrin and Sec7 domain containing 2, Pleckstrin homology and SEC7 domain-containing protein 2
Host Species Rabbit
Immunogen pleckstrin and Sec7 domain containing 2. (Uniprot# Q9BQI7)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84249
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.