missing translation for 'onlineSavingsMsg'
Learn More

PSD Antibody, Novus Biologicals™

Product Code. 18357208 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357208 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357208 Supplier Novus Biologicals Supplier No. H00005662B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PSD Polyclonal antibody specifically detects PSD in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_002770.3
Gene Alias Exchange factor for ARF6, KIAA2011Pleckstrin homology and SEC7 domain-containing protein 1, pleckstrin and Sec7 domain containing, pleckstrin and Sec7 domain protein, PSD1PH and SEC7 domain-containing protein 1, TYLEFA6
Host Species Mouse
Immunogen PSD (NP_002770.3, 1 a.a. - 1024 a.a.) full-length human protein. MAQGAMRFCSEGDCAISPPRCPRRWLPEGPVPQSPPASMYGSTGSLLRRVAGPGPRGRELGRVTAPCTPLRGPPSPRVAPSPWAPSSPTGQPPPGAQSSVVIFRFVEKASVRPLNGLPAPGGLSRSWDLGGVSPPRPTPALGPGSNRKLRLEASTSDPLPARGGSALPGSRNLVHGPPAPPQVGADGLYSSLPNGLGGPPERLATLFGGPADTGFLNQGDTWSSPREVSSHAQRIARAKWEFFYGSLDPPSSGAKPPEQAPPSPPGVGSRQGSGVAVGRAAKYSETDLDTVPLRCYRETDIDEVLAEREEADSAIESQPSSEGPPGTAYPPAPRPGPLPGPHPSLGSGNEDEDDDEAGGEEDVDDEVFEASEGARPGSRMPLKSPVPFLPGTSPSADGPDSFSCVFEAILESHRAKGTSYTSLASLEALASPGPTQSPFFTFELPPQPPAPRPDPPAPAPLAPLEPDSGTSSAADGPWTQRGEEEEAEARAKLAPGREPPSPCHSEDSLGLGAAPLGSEPPLSQLVSDSDSELDSTERLALGSTDTLSNGQKADLEAAQRLAKRLYRLDGFRKADVARHLGKNNDFSKLVAGEYLKFFVFTGMTLDQALRVFLKELALMGETQERERVLAHFSQRYFQCNPEALSSEDGAHTLTCALMLLNTDLHGHNIGKRMTCGDFIGNLEGLNDGGDFPRELLKALYSSIKNEKLQWAIDEEELRRSLSELADPNPKVIKRISGGSGSGSSPFLDLTPEPGAAVYKHGALVRKVHADPDCRKTPRGKRGWKSFHGILKGMILYLQKEEYKPGKALSETELKNAISIHHALATRASDYSKRPHVFYLRTADWRVFLFQAPSLEQMQSWITRINVVAAMFSAPPFPAAVSSQKKFSRPLLPSAATRLSQEEQVRTHEAKLKAMASELREHRAAQLGKKGRGKEAEEQRQKEAYLEFEKSRYSTYAALLRVKLKAGSEELDAVEAALAQAGSTEDGLPPSHSSPSLQPKPSSQPRAQRHSSEPRPGAGSGRRKP
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5662
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.