Learn More
PSD-95 Antibody (6G6-1C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB300-556
Product Code. 18751714
  • £476.00 / 100µL
Description
PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSD-95
Monoclonal
3.3 mg/mL
Western Blot 1:2000, Flow Cytometry 2 ug/ test (100ul), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize
DLG4
Purified recombinant rat PSD-95.
100 μL
Neuronal Cell Markers, Neuroscience, Neurotransmission
1742
Human, Mouse, Rat, Invertebrate, Primate
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
6G6-1C9
Unconjugated
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15
Mouse
Protein A purified
RUO
Primary
Detects Post Synaptic Density 95 kDa (PSD-95) from rat tissues.
Store at -20C. Avoid freeze-thaw cycles.
IgG2a