missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSCA Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543623
This item is not returnable.
View return policy
Description
PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.
Specifications
| PSCA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O43653 | |
| PSCA | |
| Leu13-Ser86 with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| PSCA | |
| 2210408B04Rik; caldesmon; CDM; HCAD; H-CAD; LCAD; L-CAD; NAG22; PRO232; prostate stem cell antigen; PSCA; UNQ206; UNQ206/PRO232 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction