missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSCA Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21421AF750
This item is not returnable.
View return policy
Description
PSCA Polyclonal antibody specifically detects PSCA in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PSCA | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| PRO232, prostate stem cell antigen | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSCA | |
| 0.1 mL | |
| Cancer, Cellular Markers, Prostate Cancer, Stem Cell Markers | |
| 8000 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction