missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PSB CB5

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PSB CB5 is a selective GPR18 antagonist (IC50 = 0.279 μM). Exhibits >36-fold selectivity versus CB1 and GPR55 and 14-fold selectivity versus CB2 receptors. Inhibits N-Arachidonylglycine (Cat. No. 1445) induced acrosomal exocytosis in human spermatozoa.
Specifications
Specifications
| Chemical Name or Material | (2 Z)-2 [[3-[(4-Chlorophenyl)methoxy]phenyl]methylene]-6,7-dihydro-5 H-imidazo[2,1-b][1,3 ]thiazin-3(2 H)-one |
| CAS | 1627710-30-8 |
| Quantity | 50 mg |
| Target | Additional Cannabinoid-related Compounds |
| Molecular Formula | C20H17ClN2O2S |
| Formula Weight | 8030 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction