missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PSB 603

Description
Adenosine A2B receptor antagonist that displays > 17000-fold selectivity over other adenosine receptors (Ki values are 0.553, > 10000, > 10000 and > 10000 nM for A2B, A1, A2A and A3 receptors respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 529.01 |
| Purity | >98% |
| Molecular Weight (g/mol) | 526.996 |
| Chemical Name or Material | PSB 603 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?