missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSB-12062, MedChemExpress
Description
PSB-12062 is a potent and selective P2X4 antagonist with an IC50 of 1.38 μM for human P2X4.
Specifications
Specifications
| Chemical Name or Material | PSB-12062 |
| CAS | 55476-47-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H15NO3S |
| Quantity | 100 mg |
| Synonym | N-(p-Methylphenylsulfonyl)phenoxazine |
| Solubility Information | DMSO : 25 mg/mL (74.10 mM; Need ultrasonic) |
| SMILES | O=S(N1C2=C(C=CC=C2)OC3=CC=CC=C13)(C4=CC=C(C)C=C4)=O |
| Molecular Weight (g/mol) | 337.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?