missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PSB 1115

Product Code. 18025682
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18025682 50 mg 50mg
18035492 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025682 Supplier R&D Systems™ Supplier No. 2009/50
  • ONLINE EXCLUSIVE PRICE
    £708.42
    £635.25 / 50mg
    Save £73.17
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective human A2B receptor antagonist; water-soluble

Highly selective, water-soluble, human A2B adenosine receptor antagonist. Ki values are 53.4, > 10000 and > 10000 nM at human A2B, A1 and A3 receptors respectively. Also selective versus rat A1 and A2A receptors (Ki values are 2200 and 24000 nM respectively). Produces potent analgesic effects in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152529-79-8
Molecular Formula C14H14N4O5S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 350.35
Purity >98%
Molecular Weight (g/mol) 350.349
Chemical Name or Material PSB 1115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.