missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PSB 0739

Product Code. 18064152
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18064152 10 mg 10mg
18005852 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064152 Supplier Tocris Bioscience™ Supplier No. 3983/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent P2Y12 antagonist

Highly potent P2Y12 receptor antagonist (Ki = 24.9 nM). Unlike clopidogrel (Cat. No. 2490), does not require bioactivation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1052087-90-7
Molecular Formula C26H19N3Na2O8S2

Specifications

Quantity 10 mg
Formula Weight 609.54
Purity >96%
Molecular Weight (g/mol) 611.551
Chemical Name or Material PSB 0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.