missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSAT1 Recombinant Rabbit Monoclonal Antibody (HL2663)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556849
This item is not returnable.
View return policy
Description
PSAT1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
| PSAT1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q99K85, Q9Y617 | |
| PSAT1 | |
| Recombinant fragment of human PSAT1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2663 | |
| Unconjugated | |
| PSAT1 | |
| D8Ertd814e; endometrial progesterone-induced protein; EPIP; NLS2; Phosphohydroxythreonine aminotransferase; phosphoserine aminotransferase; phosphoserine aminotransferase 1; PSA; Psa1; Psat; Psat1; PSATD | |
| Rabbit | |
| Protein A | |
| RUO | |
| 107272, 293820, 29968 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction