Learn More
PSAT1 Rabbit, anti-Human, CoraLite Plus 488, Clone: 1O22, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | PSAT1 |
|---|---|
| Clone | 1O22 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18080736
|
Proteintech
CL488-81479-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088065
|
Proteintech
CL488-81479-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PSAT1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PSAT1. This antibody recognizes human, mouse, pig antigen. The PSAT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PSAT1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 29968 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1O22 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Pig | |
| EPIP, Phosphoserine aminotransferase, PSA, PSAT, PSAT1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.