missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121666
This item is not returnable.
View return policy
Description
PSAT1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
| PSAT1 | |
| Polyclonal | |
| Unconjugated | |
| PSAT1 | |
| D8Ertd814e; endometrial progesterone-induced protein; EPIP; NLS2; Phosphohydroxythreonine aminotransferase; phosphoserine aminotransferase; phosphoserine aminotransferase 1; PSA; Psa1; Psat; Psat1; PSATD | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100154160, 107272, 293820, 29968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q99K85, Q9Y617 | |
| PSAT1 | |
| Recombinant protein Phosphoserine Aminotransferase 1. The antigen corresponds to amino acid range 1-312 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction