missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PSAT1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243154
|
Novus Biologicals
NBP2-55269 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624437
|
Novus Biologicals
NBP2-55269-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSAT1 Polyclonal specifically detects PSAT1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PSAT1 | |
| Polyclonal | |
| Rabbit | |
| metabolism | |
| EC 2.6.1.52, EPIP, Phosphohydroxythreonine aminotransferase, phosphoserine aminotransferase, phosphoserine aminotransferase 1, PSAMGC1460, PSATendometrial progesterone-induced protein | |
| PSAT1 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 29968 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GAWSAKAAEEAKKFGTINIVHPKLGSYTKIPDPSTWNLNPDASYVYYCANETVHGVEFDFIPDVKGAVLVCDMSSNFLSKPVDVSKFGVIFAGAQKNVGSAG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title