Learn More
Invitrogen™ PSAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110226
Description
Immunogen sequence: KEMPMQTLVP AKVASKNVIP ALELVEPIKK HEVPAKSDVY CEVCEFLVKE VTKLIDNNKT EKEILDAFDK MCSKLPKSLS EECQEVVDTY GSSILSILLE EVSPELVCSM LHLCSGTRLP ALTVHVTQPK DGGFCEVCKK LVGYLDRNLE KNSTKQEILA ALEKGCSFLP DPYQKQCDQF VAEYEPVLIE ILVEVMDPSF VCLKIGACPS AHKPLLGTEK CIWGPSYWCQ NTETAAQCNA VEHCKRHVWN.
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
Specifications
| PSAP | |
| Polyclonal | |
| Unconjugated | |
| Psap | |
| A1 activator; AI037048; Cerebroside sulfate activator; Co-beta-glucosidase; Component C; CSAct; Dispersin; GLBA; Glucosylceramidase activator; Proactivator polypeptide; prosaposin; prosaposin (sulfated glycoprotein, sphingolipid hydrolase activator); Protein A; Protein C; Psap; SAP1; SAP-1; SAP-2; Saposin-A; Saposin-B; Saposin-B-Val; Saposin-C; Saposin-D; Sgp1; SGP-1; SGP1A; snoRNA MBII-198; Sphingolipid activator protein 1; Sphingolipid activator protein 2; sphingolipid activator protein-1; sulfated glycoprotein 1; Sulfatide/GM1 activator | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19156, 25524, 5660 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P07602, P10960, Q61207 | |
| Psap | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.