missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAP Mouse anti-Human, Clone: 2F11B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66473-1-IG-20UL
This item is not returnable.
View return policy
Description
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.Specifications
| PSAP | |
| Monoclonal | |
| 0.99 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07602 | |
| PSAP | |
| PSAP Fusion Protein ag1194 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2F11B2 | |
| Unconjugated | |
| PSAP | |
| A1 activator, Cerebroside sulfate activator, Co beta glucosidase, Component C, CSAct, Dispersin, GLBA, Glucosylceramidase activator, Proactivator polypeptide, prosaposin, Protein C, PSAP, SAP 1, SAP 2 Saposin D, SAP1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5660 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction