Learn More
PSA Monoclonal Antibody (EP1588Y), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514470
Description
MA5-14470 targets Prostate Specific Antigen in IHC (P), ICC/IF and WB applications and shows reactivity with Human samples, however this antibody is not recommended for human colon and mouse prostate tissue sections. This antibody does not react with rat tissue in Western blot applications. The MA5-14470 immunogen is a synthetic peptide corresponding to residues near the C-terminus of human PSA.
PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.
Specifications
| PSA | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 15mM sodium azide | |
| P07288 | |
| KLK3 | |
| A synthetic peptide corresponding to residues near the C-terminus of human PSA | |
| RUO | |
| 354 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| EP1588Y | |
| Unconjugated | |
| KLK3 | |
| Antigen; antigen, prostate specific; APS; Fletcher factor; gamma-seminoprotein; hK3; Kal3; Kal-3; Kallikrein 3; kallikrein 3, plasma; kallikrein B, plasma 1; kallikrein related peptidase 3; kallikrein-3; Kallikrein-3 (KLK3); kallikrein-related peptidase 3; Kalp1; KALP15; Kininogenin; KLK2A1; Klk3; KLK-3; Klkb1; P-30 antigen; Pk; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; prostate specific antigen; prostate-specific (APS); prostate-specific antigen; PSA; Semenogelase; seminin | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.