missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Prussian Blue, pure

Product Code. p-188834 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10722451

Brand: Thermo Scientific Chemicals 215210250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14038-43-8 | C18Fe7N18 | 859.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14038-43-8
Molecular Formula C18Fe7N18
Molecular Weight (g/mol) 859.239
MDL Number MFCD00135663
InChI Key DCYOBGZUOMKFPA-UHFFFAOYSA-N
Synonym C.I. 77510, Ferric ferrocyanide, Pigment Blue 27
PubChem CID 2724251
IUPAC Name iron(2+);iron(3+);octadecacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3]

Specifications

Linear Formula Fe4[Fe(CN)6]3
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in diluted acids, practically insoluble in most common organic, solvents
Infrared Spectrum Authentic
Formula Weight 859.25
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Blue
Quantity 25 g
Physical Form Powder
Chemical Name or Material Prussian Blue
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.