missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRUNE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556632
This item is not returnable.
View return policy
Description
Immunogen sequence: MESEKISEKQ EEILSILEEK FPNLPPREDI INVLQETQFS AQGLSIEQTM LKDLKELSDG EIKVAISTVS MNLEVRVGML Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.
PRUNE2 gene, also named BMCC1, encodes protein prune homolog 2. PRUNE2 gene are overlapping with PCA3, one of the most prostate cancer specific markers. PRUNE2 may play an important role in regulating differentiation, survival and aggressiveness of the tumor cells. Besides its high level of expression seen in the nervous system (brain, cerebellum and spinal cord), it is also expressed at high levels in noneuroblastoma, rhabdomyosarcoma, melanoma and some osteosarcoma cell lines, whereas at only low levels in cancer cell lines of liver, breast, thyroid and colon. The Calculated molecular weight of this protein is 340 kDa.
Specifications
| PRUNE2 | |
| Polyclonal | |
| Unconjugated | |
| PRUNE2 | |
| 6330414G02Rik; A214N16.3; A230083H22Rik; A330102H22Rik; bA214N16.3; BCH motif-containing molecule at the carboxyl terminal region 1; Bmcc1; BNIP2 motif containing molecule at the carboxyl terminal region 1; BNIP2 motif-containing molecule at the C-terminal region 1; BNIPXL; C9orf65; DKFZp762K117; KIAA0367; mKIAA0367; neuronal protein; olfaxin; protein prune homolog 2; prune homolog 2; prune homolog 2 (Drosophila); PRUNE2; RGD1311350; RP11-214N16.3; RP11-58J3.2; truncated PRUNE2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158471 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUY3 | |
| PRUNE2 | |
| Recombinant protein corresponding to Human PRUNE2. Recombinant protein control fragment (Product #RP-93020). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction