missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRUNE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | PRUNE2 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18493811
|
Novus Biologicals
NBP1-89336-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18293656
|
Novus Biologicals
NBP1-89336 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRUNE2 Polyclonal specifically detects PRUNE2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PRUNE2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 158471 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LNDEGKLSITLVGSSVLASEDKTLESAVVKVINPVEQSDANVEFRESSSSLVLKEILQEAPELITEQLAHRLRGSILFKWMT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| A214N16.3, bA214N16.3, BCH motif-containing molecule at the carboxyl terminal region 1, BMCC1FLJ50060, BNIP2 motif containing molecule at the carboxyl terminal region 1, BNIP2 motif-containing molecule at the C-terminal region 1, BNIPXLFLJ59118, C9orf65, chromosome 9 open reading frame 65, DKFZp762K117, KIAA0367FLJ54876, protein prune homolog 2, prune homolog 2 (Drosophila), RP11-58J3.2 | |
| PRUNE2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title