missing translation for 'onlineSavingsMsg'
Learn More

Prulifloxacin, TRC

Product Code. 30428786
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428786 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428786 Supplier Toronto Research Chemicals Supplier No. TRCP838885250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prulifloxacin
CAS 123447-62-1
Purity Grade Notes HPLC
Molecular Formula C21 H20 F N3 O6 S
Quantity 250 mg
Synonym 1H,4H-[1,3]Thiazeto[3,2-a]quinoline-3-carboxylic acid','6-fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]-1-piperazinyl]-4-oxo-, 1,3-Dioxole','1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid deriv., 6-Fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]-1-piperazinyl]-4-oxo-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid, (±)-7-[4-[(Z)-2,3-Dihydroxy-2-butenyl]-1-piperazinyl]-6-fluoro-1-methyl-4-oxo-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid','cyclic carbonate, NM 441, Prulifloxacin, Quisnon, Sword
InChI Formula InChI=1S/C21H20FN3O6S/c1-10-16(31-21(29)30-10)9-23-3-5-24(6-4-23)15-8-14-12(7-13(15)22)18(26)17(20(27)28)19-25(14)11(2)32-19/h7-8,11H,3-6,9H2,1-2H3,(H,27,28)
SMILES CC1SC2=C(C(=O)O)C(=O)c3cc(F)c(cc3N12)N4CCN(CC5=C(C)OC(=O)O5)CC4
IUPAC Name 6-fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]piperazin-1-yl]-4-oxo-1H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
Molecular Weight (g/mol) 461.46
Formula Weight 461.1057
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.