missing translation for 'onlineSavingsMsg'
Learn More

Prulifloxacin, MedChemExpress

Product Code. 16461996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461996 500 mg 500mg
16448066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461996 Supplier Medchem Express Supplier No. HYB0024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Prulifloxacin (NM441) is an orally active fluoroquinolone antibiotic with a broad spectrum of activity against Gram-positive and -negative bacteria. Prulifloxacin is a prodrug of a thiazeto-quinoline carboxylic acid derivative Ulifloxacin (NM394). Prulifloxacin has the potential for lower urinary tract infections and exacerbations of chronic bronchitis.

Prulifloxacin (NM441) is an orally active fluoroquinolone antibiotic with a broad spectrum of activity against Gram-positive and -negative bacteria. Prulifloxacin is a prodrug of a thiazeto-quinoline carboxylic acid derivative Ulifloxacin (NM394). Prulifloxacin has the potential for lower urinary tract infections and exacerbations of chronic bronchitis.

Specifications

Chemical Name or Material Prulifloxacin
Physical Form Solid
CAS 123447-62-1
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C21H20FN3O6S
For Use With (Application) COVID-19-immunoregulation
Synonym NM441
Solubility Information DMSO : 12.5 mg/mL (27.09 mM; Need ultrasonic)
SMILES O=C(C1=C(SC2C)N2C3=C(C=C(F)C(N4CCN(CC5=C(C)OC(O5)=O)CC4)=C3)C1=O)O
Molecular Weight (g/mol) 461.46
Formula Weight 461.46
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.