missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRTFDC1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00245CL1
This item is not returnable.
View return policy
Description
PRTFDC1 Polyclonal antibody specifically detects PRTFDC1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| PRTFDC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| EC 2.4.2.8, FLJ11888, HHGPEC 2.4.2, phosphoribosyl transferase domain containing 1, phosphoribosyltransferase domain-containing protein 1 | |
| Produced in rabbits immunized with purified, recombinant Human PRTFDC1 (Uniprot#: Q9NRG1-1; Met1-Val225) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 56952 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction